Skip to Content
Merck
All Photos(1)

Documents

W304700

Sigma-Aldrich

Terpinyl acetate

≥95% (mixture of isomers)

Synonym(s):

(±)-α-Terpinyl acetate, predominantly α-isomer, (±)-2-(4-Methyl-3-cyclohexenyl)isopropyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20O2
CAS Number:
Molecular Weight:
196.29
FEMA Number:
3047
Beilstein:
3198769
EC Number:
Council of Europe no.:
205
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.015

biological source

synthetic

grade

Halal
Kosher

reg. compliance

FDA 21 CFR 172.515

Assay

≥95% (mixture of isomers)

refractive index

n20/D 1.465 (lit.)

bp

220 °C (lit.)

density

0.953 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

herbaceous; citrus; spicy

SMILES string

CC(=O)OC(C)(C)C1CCC(C)=CC1

InChI

1S/C12H20O2/c1-9-5-7-11(8-6-9)12(3,4)14-10(2)13/h5,11H,6-8H2,1-4H3

InChI key

IGODOXYLBBXFDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valeria Cagno et al.
PloS one, 12(2), e0172322-e0172322 (2017-02-17)
Salvia desoleana Atzei & V. Picci is an indigenous species in Sardinia island used in folk medicine to treat menstrual, digestive and central nervous system diseases. Nowadays, it is widely cultivated for the pleasant smell of its essential oil (EO)
Syed Ibrahim Shah et al.
Toxicology in vitro : an international journal published in association with BIBRA, 69, 105004-105004 (2020-10-04)
Here we report a new planarian (Dugesia lugubris) fluorescent assay as a rapid and cheap pre-screening tool to predict strong skin irritants. Our aim was to provide a simple and cost-effective in vivo method that avoided use of higher vertebrates.
Nicholas J Sadgrove et al.
Plants (Basel, Switzerland), 9(11) (2020-11-19)
The highly aromatic Australian mint bushes from the genus Prostanthera Labill. produce a high yield of essential oil on hydrodistillation. Together with its rich history, horticultural potential, iconic flowers, and aromatic leaves, it achieves high ornamental and culinary value. Species
Wei-Na Kong et al.
Pest management science, 76(9), 3225-3235 (2020-05-02)
Grapholita molesta (Busck) is a fruit pest worldwide. While sex pheromones-based technology for male attraction has made great progress in the monitoring or mass trapping of G. molesta, the attraction of males alone is not an effective reproductive control strategy.
Alexis Marsol-Vall et al.
Food chemistry, 293, 151-160 (2019-06-04)
Blackcurrant juices were prepared without enzyme (NEB-juice) and with conventional enzyme-aided berry pressing (EB-juice). Juices were pasteurized and stored at ambient temperature (in light and dark conditions) and at +4 °C for a 1-year period of time. Volatile composition and odor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service