Skip to Content
Merck
All Photos(1)

Key Documents

W297607

Sigma-Aldrich

Quinine monohydrochloride dihydrate

≥95%, FG

Synonym(s):

Quinine monohydrochloride dihydrate, Quinine hydrochloride dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

W297607-SAMPLE-K
€48.80
100 G
€60.60
1 KG
€383.00
5 KG
€2,100.00

€48.80


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W297607-SAMPLE-K
€48.80
100 G
€60.60
1 KG
€383.00
5 KG
€2,100.00

About This Item

Empirical Formula (Hill Notation):
C20H24N2O2 · HCl · 2H2O
CAS Number:
Molecular Weight:
396.91
FEMA Number:
2976
Beilstein:
6112655
EC Number:
Council of Europe no.:
715
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.155
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

€48.80


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

Assay

≥95%

optical activity

[α]22/D −235°, c = 2 in 0.1 M HCl

mp

115-116 °C (dec.) (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

Cl[H].[H]O[H].[H]O[H].[H][C@@]12CCN(C[C@@H]1C=C)[C@@]([H])(C2)[C@H](O)c3ccnc4ccc(OC)cc34

InChI

1S/C20H24N2O2.ClH.2H2O/c1-3-13-12-22-9-7-14(13)10-19(22)20(23)16-6-8-21-18-5-4-15(24-2)11-17(16)18;;;/h3-6,8,11,13-14,19-20,23H,1,7,9-10,12H2,2H3;1H;2*1H2/t13-,14-,19-,20+;;;/m0.../s1

InChI key

MPQKYZPYCSTMEI-FLZPLBAKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Rats can predict aversiveness of Active Pharmaceutical Ingredients.: This study explores the use of rats to predict the aversiveness of various active pharmaceutical ingredients (APIs), including Quinine hydrochloride dihydrate, by measuring behavioral responses. The findings suggest that animal models can be effective in preclinical assessments of API aversiveness, which is crucial for drug formulation and patient compliance (Soto et al., 2018).

  • Impact of sodium lauryl sulfate in oral liquids on e-tongue measurements.: This research investigates the influence of sodium lauryl sulfate on electronic tongue measurements of bitterness in oral liquid formulations containing Quinine hydrochloride dihydrate. The study highlights the importance of excipients in modifying taste perception, which is significant for developing palatable pharmaceutical products (Immohr et al., 2016).

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service