Skip to Content
Merck
All Photos(1)

Documents

W292451

Sigma-Aldrich

Propionic acid

feed grade, 98.7%

Synonym(s):

Acid C3, Propanoic acid, Propanyl acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COOH
CAS Number:
Molecular Weight:
74.08
FEMA Number:
2924
Beilstein:
506071
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.003

biological source

synthetic

grade

feed grade

vapor density

2.55 (vs air)

vapor pressure

2.4 mmHg ( 20 °C)

Assay

98.7%

autoignition temp.

955 °F

expl. lim.

12.1 %

refractive index

n20/D 1.386 (lit.)
n20/D 1.386

bp

141 °C (lit.)
141 °C

mp

−24-−23 °C (lit.)

density

0.993 g/mL at 25 °C (lit.)
0.995 g/mL at 25 °C

application(s)

flavors and fragrances

Organoleptic

vinegar

SMILES string

CCC(O)=O

InChI

1S/C3H6O2/c1-2-3(4)5/h2H2,1H3,(H,4,5)

InChI key

XBDQKXXYIPTUBI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.

Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R De Kanter et al.
Xenobiotica; the fate of foreign compounds in biological systems, 32(5), 349-362 (2002-06-18)
1. Organ-specific biotransformation was studied in human and rat liver, lung, kidney and small intestine slices and compared on a protein basis, using four model substances. 2. Deethylation of lidocaine was highest in liver slices from both man and rat
Weiqiang Chen et al.
Journal of virology, 89(1), 581-593 (2014-10-24)
The recent global resurgence of arthritogenic alphaviruses, in particular chikungunya virus (CHIKV), highlights an urgent need for the development of therapeutic intervention strategies. While there has been significant progress in defining the pathophysiology of alphaviral disease, relatively little is known
Sa'ad H Al-Lahham et al.
Biochimica et biophysica acta, 1801(11), 1175-1183 (2010-08-10)
Undigested food is fermented in the colon by the microbiota and gives rise to various microbial metabolites. Short-chain fatty acids (SCFA), including acetic, propionic and butyric acid, are the principal metabolites produced. However, most of the literature focuses on butyrate
L B Bindels et al.
British journal of cancer, 107(8), 1337-1344 (2012-09-15)
Metabolites released by the gut microbiota may influence host metabolism and immunity. We have tested the hypothesis that inulin-type fructans (ITF), by promoting microbial production of short-chain fatty acids (SCFA), influence cancer cell proliferation outside the gut. Mice transplanted with
S Somerma et al.
Acta dermato-venereologica, 64(1), 41-45 (1984-01-01)
The effect of five corticosteroid ointments on epidermal thickness was studied using occlusive chamber application. Thinning of the epidermis and telangiectasia were noted 3 weeks after application of betamethasone-17-valerate (BV), budesonide (BD), clobetasol-17-propionate (CP), and fluocinolone acetonide (FA), but not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service