Skip to Content
Merck
All Photos(1)

Key Documents

W287318

Sigma-Aldrich

2-Phenoxyethyl isobutyrate

≥97%, FCC

Synonym(s):

Ethylene glycol monophenylether isobutyrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCO2CH2CH2OC6H5
CAS Number:
Molecular Weight:
208.25
FEMA Number:
2873
EC Number:
Council of Europe no.:
2089
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.487

biological source

synthetic

reg. compliance

FCC
FDA 21 CFR 172.515

Assay

≥97%

refractive index

n20/D 1.493 (lit.)

bp

125-127 °C/4 mmHg (lit.)

density

1.044 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

honey; rose

SMILES string

CC(C)C(=O)OCCOc1ccccc1

InChI

1S/C12H16O3/c1-10(2)12(13)15-9-8-14-11-6-4-3-5-7-11/h3-7,10H,8-9H2,1-2H3

InChI key

MJTPMXWJHPOWGH-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

278.9 °F - closed cup

Flash Point(C)

137.16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service