Skip to Content
Merck
All Photos(1)

Documents

W270027

Sigma-Aldrich

3-Methyl-1,2-cyclopentanedione

98%

Synonym(s):

2-Hydroxy-3-methyl-2-cyclopenten-1-one, Maple lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O2
CAS Number:
Molecular Weight:
112.13
FEMA Number:
2700
Beilstein:
2076520
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:

biological source

synthetic

Quality Level

Assay

98%

mp

104-108 °C (lit.)
104-108 °C

application(s)

flavors and fragrances

SMILES string

CC1CCC(=O)C1=O

InChI

1S/C6H8O2/c1-4-2-3-5(7)6(4)8/h4H,2-3H2,1H3

InChI key

OACYKCIZDVVNJL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seo Young Choi et al.
Biochimica et biophysica acta, 1770(12), 1612-1619 (2007-10-13)
Peroxisome proliferator-activated receptors (PPARs) are members of the nuclear hormone receptor superfamily that plays a pivotal role in regulating inflammatory gene expression. The purpose of this study was to investigate the effects of coffee extract, 3-methyl-1,2-cyclopentanedione (3-MCP) on PPARs in
Jae Heun Chung et al.
Journal of agricultural and food chemistry, 55(16), 6787-6792 (2007-07-17)
Activation of the redox-sensitive transcription factor, nuclear factor-kappa B (NF-kappaB), plays a central role in inflammation and aging processes by inducing pro-inflammatory genes. The present study was designed to unravel the molecular mechanisms underlying the anti-inflammation effects of 3-methyl-1,2-cyclopentanedione (3-MCP)
Ae Ra Kim et al.
The Journal of pharmacy and pharmacology, 54(10), 1385-1392 (2002-10-25)
It has been known that reactive oxygen and nitrogen species such as nitric oxide (NO), superoxide radical (*O2-) and their byproduct peroxynitrite (ONOO-) induce cellular and tissue injury, ultimately resulting in several human diseases. In this study, we examined scavenging
Bryson W Katona et al.
The Journal of organic chemistry, 72(24), 9298-9307 (2007-10-26)
Deoxycholic acid (DCA) is an endogenous secondary bile acid implicated in numerous pathological conditions including colon cancer formation and progression and cholestatic liver disease. DCA involvement in these disease processes results partly from its ability to modulate signaling cascades within
Bruna R Toledo et al.
Journal of chromatography. A, 1346, 1-7 (2014-05-03)
Solid-phase microextraction (SPME) using cross-linked polymeric ionic liquid (PIL)-based sorbent coatings was used to extract volatile aroma-related compounds from coffee samples. Several PIL-based coatings were screened alongside a commercial poly(acrylate) (PA) SPME coating. The best performing PIL-based SPME fiber, poly(1-vinyl-3-hexadecylimidazolium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service