Skip to Content
Merck
All Photos(1)

Documents

W262528

Sigma-Aldrich

Lemon oil

cold-pressed, California origin, FG

Synonym(s):

Lemon oil c.p. california

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
FEMA Number:
2625
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21

biological source

Citrus limon

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117
FDA 21 CFR 182.20

optical activity

[α]20/D +64°, neat

quality

cold-pressed

composition

may contain IFRA restricted Citral, ≤3.0%
may contain IFRA restricted Geraniol, ≤0.2%

origin

California origin

refractive index

n20/D 1.473 (lit.)

bp

176 °C (lit.)

density

0.850 g/mL at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

lemon; citrus

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Preparation, Characterization and Biological Activities of an Oil-in-Water Nanoemulsion from Fish By-Products and Lemon Oil by Ultrasonication Method.: This paper presents the preparation and biological evaluation of a nanoemulsion combining fish by-products and lemon oil. The study highlights the nanoemulsion′s potential antimicrobial and antioxidant activities (Azmi NAN et al., 2022).

  • Impact of pea protein-inulin conjugates prepared via the Maillard reaction using a combination of ultrasound and pH-shift treatments on physical and oxidative stability of algae oil emulsions.: This research examines the impact of pea protein-inulin conjugates on the stability of algae oil emulsions, with lemon oil used to assess oxidative stability improvements (Jiang W et al., 2022).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaun Yon-Seng Khoo et al.
Frontiers in behavioral neuroscience, 14, 5-5 (2020-03-03)
Conditioned responding can be renewed by re-exposure to the conditioning context following extinction in a different context (ABA renewal) or by removal from the extinction context (AAB or ABC renewal). ABA renewal is robust in Pavlovian and operant conditioning paradigms.
M Flavia Barbano et al.
Neuron, 107(2), 368-382 (2020-05-23)
The ventral tegmental area (VTA) has dopamine, GABA, and glutamate neurons, which have been implicated in reward and aversion. Here, we determined whether VTA-glutamate or -GABA neurons play a role in innate defensive behavior. By VTA cell-type-specific genetic ablation, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service