Skip to Content
Merck
All Photos(1)

Key Documents

W248835

Sigma-Aldrich

Fumaric acid

natural (US), ≥98%, FG

Sign Into View Organizational & Contract Pricing

Select a Size

W248835-SAMPLE-K
€52.30
1 KG
€157.00
5 KG
€550.00

€52.30


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W248835-SAMPLE-K
€52.30
1 KG
€157.00
5 KG
€550.00

About This Item

Linear Formula:
HOOCCH=CHCOOH
CAS Number:
Molecular Weight:
116.07
FEMA Number:
2488
Beilstein:
605763
Council of Europe no.:
25
MDL number:
UNSPSC Code:
12164502
Flavis number:
08.025
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural (US)
biological source:
botanical
food allergen:
no known allergens

€52.30


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

biological source

botanical

Quality Level

grade

FG
Halal
Kosher
natural (US)

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 117

vapor pressure

1.7 mmHg ( 165 °C)

Assay

≥98%

autoignition temp.

1364 °F

expl. lim.

40 %

mp

298-300 °C (subl.) (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

OC(=O)\C=C\C(O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1+

InChI key

VZCYOOQTPOCHFL-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

523.4 °F

Flash Point(C)

273 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service