Skip to Content
Merck
All Photos(1)

Key Documents

W245402

Sigma-Aldrich

Ethyl 3-phenylglycidate

mixture of cis and trans, 92%

Sign Into View Organizational & Contract Pricing

Select a Size

W245402-SAMPLE
€47.40
1 KG
€124.00
5 KG
€474.00

€47.40


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W245402-SAMPLE
€47.40
1 KG
€124.00
5 KG
€474.00

About This Item

Empirical Formula (Hill Notation):
C11H12O3
CAS Number:
Molecular Weight:
192.21
FEMA Number:
2454
EC Number:
Council of Europe no.:
11844
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
16.018
NACRES:
NA.21
grade:
Halal
biological source:
synthetic
food allergen:
no known allergens

€47.40


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

300
400

grade

Halal

reg. compliance

FDA 21 CFR 172.515

Assay

92%

refractive index

n20/D 1.518 (lit.)

bp

96 °C/0.5 mmHg (lit.)

density

1.102 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

berry; floral; strawberry; sweet

SMILES string

CCOC(=O)C1OC1c2ccccc2

InChI

1S/C11H12O3/c1-2-13-11(12)10-9(14-10)8-6-4-3-5-7-8/h3-7,9-10H,2H2,1H3

InChI key

GOMAKLPNAAZVCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Congfa Li et al.
Biotechnology letters, 25(24), 2113-2116 (2004-02-19)
A Pseudomonas sp. was isolated with enantioselective epoxide hydrolase activity to ethyl 3-phenylglycidate. Cells grown on sucrose and suspended in 10% (v/v) dimethyl formamide as co-solvent produced (2R,3S) ethyl 3-phenylglycidate with 95% ee and 26% yield in 12 h from
C Tilch et al.
Mutation research, 138(1), 1-8 (1984-10-01)
EPG and an in vitro digest of EPG by pepsin and pancreatin simulating mammalian digestion have been examined for genotoxicity in 4 mutagenicity tests employing different genetic endpoints. In the Salmonella reverse mutation assay, EPG showed only slight mutagenic activity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service