Skip to Content
Merck
All Photos(1)

Key Documents

W217808

Sigma-Aldrich

Butyl alcohol

≥99.9%, FCC

Synonym(s):

1-Butanol, n-Butanol, Butyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OH
CAS Number:
Molecular Weight:
74.12
FEMA Number:
2178
Beilstein:
969148
EC Number:
Council of Europe no.:
52
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.004

biological source

synthetic

grade

Kosher

reg. compliance

FCC
FDA 21 CFR 172.515

vapor density

2.55 (vs air)

Assay

≥99.9%

autoignition temp.

649 °F

expl. lim.

11.2 %

refractive index

n20/D 1.399 (lit.)

bp

116-118 °C (lit.)

mp

−90 °C (lit.)

density

0.81 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

banana; vanilla

SMILES string

CCCCO

InChI

1S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3

InChI key

LRHPLDYGYMQRHN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - Pensky-Martens closed cup

Flash Point(C)

35 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R W Wulkan et al.
The International journal of biochemistry, 18(11), 1045-1051 (1986-01-01)
The butanol extraction method of Morton (1950), a routine step in enzyme purification, is discussed with special reference to a hydrophobic form of alkaline phosphatase from human liver tissue. This form slowly precipitates from butanol-extracted liver tissue homogenates stored at
Joel G Davis et al.
Nature, 491(7425), 582-585 (2012-11-23)
Hydrophobic hydration is considered to have a key role in biological processes ranging from membrane formation to protein folding and ligand binding. Historically, hydrophobic hydration shells were thought to resemble solid clathrate hydrates, with solutes surrounded by polyhedral cages composed
A L Gainer et al.
Clinica chimica acta; international journal of clinical chemistry, 123(1-2), 11-17 (1982-08-04)
Neutrophils were isolated in good yield from fresh whole blood and their alkaline phosphatase was solubilized. Inhibitor studies using L-phenylalanylglycylglycine, L-phenylalanine and L-homoarginine revealed a distinct pattern of inhibition for each of the crude or purified preparations of the human
Kazuhiro Yamamoto et al.
European heart journal, 25(14), 1221-1229 (2004-07-13)
Ventricular fibrosis is promoted by many effectors that chronically activate phospholipase D (PLD), and induces cardiac dysfunction and heart failure in cardiovascular diseases. Since ethanolamine is a product of PLD, we hypothesised that an administration of an analogue of ethanolamine
Jie Yao et al.
Nature communications, 5, 5670-5670 (2014-11-29)
Layer-structured two-dimensional nanomaterials are a family of materials with strong covalent bonding within layers and weak van der Waals interaction between layers, whose vertical thickness can be thinned down to few nanometer and even single atomic layer. Bismuth chalcogenides are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service