Skip to Content
Merck
All Photos(1)

Documents

W208302

Sigma-Aldrich

Isoamyl pyruvate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCO2CH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
158.19
FEMA Number:
2083
EC Number:
Council of Europe no.:
431
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.443

biological source

synthetic

reg. compliance

FDA 21 CFR 172.515

refractive index

n20/D 1.419 (lit.)

bp

185 °C (lit.)

density

0.978 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Organoleptic

balsamic

SMILES string

CC(C)CCOC(=O)C(C)=O

InChI

1S/C8H14O3/c1-6(2)4-5-11-8(10)7(3)9/h6H,4-5H2,1-3H3

InChI key

LBKWGGFVEDOVEP-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service