Key Documents
TEM-74357
Sigma-Aldrich
Copper Tomography Grid R14C2
pack of 50 ×
Synonym(s):
Copper Tomography Grid, 300 mesh, 50pk
About This Item
UNSPSC Code:
41111761
NACRES:
MA.24
Product TEM-74357 is not currently sold in your country. Contact Technical Service
packaging
pack of 50 ×
General description
This 1.5mm square, 300 mesh grid is made from copper and is for use with Fischione tomography specimen holders. The size allows for further tilt in TEMs with small pole piece holes. An identification mark and the square shape give easy reference when rotated past 90°.
Choose from one of the most recent versions:
Certificates of Analysis (COA)