T33553
Thiourea
99%
Synonym(s):
Sulfourea, Thiocarbamide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
99%
mp
170-176 °C (lit.)
SMILES string
NC(N)=S
InChI
1S/CH4N2S/c2-1(3)4/h(H4,2,3,4)
InChI key
UMGDCJDMYOKAJW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Chaotropic agent; strong denaturant. Increases solubility and recovery of proteins
replaced by
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Repr. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Clinical & experimental metastasis, 30(7), 891-902 (2013-06-12)
Metastasis remains the major cause of therapeutic failure, poor prognosis and high mortality in breast and prostate cancer patients. Aberrant microenvironments including hypoxia and acidic pH are common features of most solid tumors that have been long associated with enhanced
Macromolecular rapid communications, 34(8), 652-658 (2013-03-14)
Readily water-soluble PEGylated amphiphiles containing bis-thiourea-based molecular recognition units at the interface of hydrophobic and hydrophilic blocks are developed. Self-assembly of these amphiphiles is found to be dependent on the exact chemical composition of the hydrophobic component. Elongated, spherical, and
The Journal of organic chemistry, 78(8), 3980-3988 (2013-03-09)
Proof that sulfur is a viable reporting element for the development of fluorescent chemosensors for metal ions is presented. To date, the majority of metal-responsive fluorescent chemosensors have relied on metal-nitrogen coordination to provide a fluorescence response, most commonly by
Chemical & pharmaceutical bulletin, 58(5), 593-601 (2010-05-13)
We have developed several multifunctional thiourea catalysts bearing a tertiary amine or an 1,2-amino alcohol in expectation of their synchronous activation of a nucleophile and an electrophile through both acid-base and hydrogen-bonding interactions. From these studies, it was revealed that
Nature chemistry, 5(12), 1049-1057 (2013-11-22)
With the intention of improving synthetic efficiency, organic chemists have turned to bioinspired organocascade or domino processes that generate multiple bonds and stereocentres in a single operation. However, despite the great importance of substituted cyclopentanes, given their prevalence in complex
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service