Skip to Content
Merck
All Photos(1)

Key Documents

T33006

Sigma-Aldrich

(Phenylthio)acetic acid

96%

Synonym(s):

(Phenylmercapto)acetic acid, S-Phenylthioglycolic acid, Thiophenoxyacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€72.00

€72.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€72.00

About This Item

Linear Formula:
C6H5SCH2COOH
CAS Number:
Molecular Weight:
168.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€72.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

96%

mp

60-63 °C (lit.)

SMILES string

OC(=O)CSc1ccccc1

InChI

1S/C8H8O2S/c9-8(10)6-11-7-4-2-1-3-5-7/h1-5H,6H2,(H,9,10)

InChI key

MOTOSAGBNXXRRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R E Saari et al.
Biochemistry, 37(9), 3035-3042 (1998-04-16)
The first step in catabolism of the broadleaf herbicide 2,4-dichlorophenoxyacetic acid (2,4-D) is catalyzed by 2,4-D/alpha-ketoglutarate (alpha-KG)-dioxygenase (TfdA) in Ralstonia eutropha (formerly Alcaligenes eutrophus) JMP134. This oxygen- and ferrous-ion-dependent enzyme couples the oxidative decarboxylation of alpha-KG (yielding CO2 and succinate)
Rui Zhang et al.
Bioorganic & medicinal chemistry letters, 17(14), 3855-3859 (2007-05-26)
A novel series of potent and selective PPARdelta agonists, para-alkylthiophenoxyacetic acids, was identified. The synthesis and structure-activity relationships are described.
A Q Zhang et al.
Chemosphere, 36(15), 3033-3041 (1998-09-25)
Batch data of aerobic microbial degradation rate constants Kb of phenylthio, phenylsulfinyl and phenylsulfonyl acetates have been determined, and the qualitative relationships between their Kb and chemical structures were analyzed. The phenylthio acetates were most subject to microbial transformation, followed
Patrick Masson et al.
Biochimica et biophysica acta, 1774(1), 16-34 (2006-12-22)
Catalytic parameters of human butyrylcholinesterase (BuChE) for hydrolysis of homologous pairs of oxo-esters and thio-esters were compared. Substrates were positively charged (benzoylcholine versus benzoylthiocholine) and neutral (phenylacetate versus phenylthioacetate). In addition to wild-type BuChE, enzymes containing mutations were used. Single
L Rodrigo et al.
Chemico-biological interactions, 119-120, 263-275 (1999-07-27)
The existence of two or more enzyme forms with paraoxonase activity has been reported in sheep, rabbit, human and rat serum and recently in mouse and rat liver. In this study we describe the presence of two peaks with paraoxonase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service