Skip to Content
Merck
All Photos(1)

Key Documents

T14605

Sigma-Aldrich

Tetrahydropyran-2-methanol

98%

Synonym(s):

2-(Hydroxymethyl)tetrahydropyran, 2-Tetrahydropyranyl methanol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€54.20

€54.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€54.20

About This Item

Empirical Formula (Hill Notation):
C6H12O2
CAS Number:
Molecular Weight:
116.16
Beilstein:
102998
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€54.20


Please contact Customer Service for Availability

Request a Bulk Order

vapor density

4.02 (vs air)

Quality Level

vapor pressure

0.4 mmHg ( 20 °C)

Assay

98%

refractive index

n20/D 1.458 (lit.)

bp

187 °C (lit.)

density

1.027 g/mL at 25 °C (lit.)

SMILES string

OCC1CCCCO1

InChI

1S/C6H12O2/c7-5-6-3-1-2-4-8-6/h6-7H,1-5H2

InChI key

ROTONRWJLXYJBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service