Skip to Content
Merck
All Photos(1)

Key Documents

T12408

Sigma-Aldrich

1,2,3,4-Tetrahydrocarbazole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H13N
CAS Number:
Molecular Weight:
171.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

bp

325-330 °C (lit.)

mp

118-120 °C (lit.)

SMILES string

C1CCc2c(C1)[nH]c3ccccc23

InChI

1S/C12H13N/c1-3-7-11-9(5-1)10-6-2-4-8-12(10)13-11/h1,3,5,7,13H,2,4,6,8H2

InChI key

XKLNOVWDVMWTOB-UHFFFAOYSA-N

Application

1,2,3,4-Tetrahydrocarbazole can be used as a starting material to prepare:
  • Spiro[cyclopentane-1,2′-indolin-3′-one] by photooxygenation.[1]
  • 9-Acyl-1,2,3,4-tetrahydrocarbazoles by N-acylation reactions.[2]
  • Carbazole via palladium-catalyzed asymmetric hydrogenation reaction.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Xu-Fan Wang et al.
Organic letters, 12(5), 1140-1143 (2010-02-12)
A hydrogen bonding-mediated double Michael addition-aromatization cascade of 2-propenylindoles and nitroolefins has been disclosed. The methodology allows an efficient synthesis of diverse and structurally complex tetrahydrocarbazoles in good to excellent enantioselectivities and diastereoselectivities.
Synthesis of novel 1-substituted and 1, 9-disubstituted-1, 2, 3, 4-tetrahydro-9H-carbazole derivatives as potential anticancer agents
Shmeiss NAMM, et al.
Molecules (Basel), 5(10), 1101-1112 (2000)
P Crawford et al.
The Journal of chemical physics, 128(10), 105104-105104 (2008-03-19)
The carbazole moiety is a component of many important pharmaceuticals including anticancer and anti-HIV agents and is commonly utilized in the production of modern polymeric materials with novel photophysical and electronic properties. Simple carbazoles are generally produced via the aromatization
Romano Di Fabio et al.
Bioorganic & medicinal chemistry letters, 16(6), 1749-1752 (2005-12-21)
The SAR of a new series of tetrahydrocarbazole derivatives is described: the appropriate decoration of this template led to the identification of a new class of NPY-1 antagonists showing good in vitro potency and a promising in vivo pharmacokinetic profile
Chao Zheng et al.
Accounts of chemical research, 53(4), 974-987 (2020-04-11)
ConspectusThe Pictet-Spengler reaction is a fundamental named reaction in organic chemistry, and it is the most straightforward method for the synthesis of tetrahydro-β-carbolines, a core structure embedded in numerous alkaloids. Spiroindolenines are often proposed as possible intermediates in Pictet-Spengler reactions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service