Key Documents
SYNS680012
Synthware low hold up high vacuum valve with metering plug
size 0-12 mm
Synonym(s):
Synthware S680012
Select a Size
1 EA
€126.00
€126.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€126.00
About This Item
UNSPSC Code:
40161501
NACRES:
NB.51
€126.00
Please contact Customer Service for Availability
packaging
pkg of 1 ea
manufacturer/tradename
Synthware S680012
Related Categories
General description
Viton® O-ring on PTFE shaft are protected with precision PTFE Wipers. The plug has a machined-in integral PTFE tip O-ring to make the primary seal. This design provides for an excellent vacuum tight seal without exposing any material other than PTFE to chemicals. With two 9mm O.D. sidearm bent to minimize hold up of liquids.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)