SYNF752475C
Synthware™ filter funnel with fritted disc
joint: ST/NS 24/40, plate O.D. 75 mm, porosity: coarse
Synonym(s):
Synthware F752475C, fritted glass funnel
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F752475C
plate O.D.
75 mm
pore size
40-60 μm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
O.D. of disc is 24 mm.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service