Key Documents
SYNC101420
Synthware Vigreux distillation column
joint: ST/NS 14/20, L 120 mm
Synonym(s):
Synthware C101420, distilling column, fractional column
Select a Size
1 EA
€66.80
€66.80
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€66.80
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
€66.80
Please contact Customer Service for Availability
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C101420
L
120 mm
Related Categories
General description
Column length is measured from the highest to lowest indent in the column.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)