Key Documents
SYNA901420
Synthware Claisen distillation adapter with compression cap thermometer port
joint: ST/NS 14/20
Synonym(s):
Synthware 901420, Glass Distillation Adapter Claisen Thermometer
Select a Size
1 EA
€79.30
€79.30
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€79.30
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
€79.30
Please contact Customer Service for Availability
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A901420
Related Categories
General description
With threaded thermometer port, supplied complete with Nylon cap and Viton® O-ring to accommodate 6-7.5 mm O.D. thermometer.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)