Key Documents
SYNA891420
Synthware distilling adapter with 105 degree bend
top joint: ST/NS 14/20, bottom joint: ST/NS 14/20
Synonym(s):
Synthware 891420, bent distillation adapter
Select a Size
1 EA
€40.50
€40.50
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€40.50
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
€40.50
Please contact Customer Service for Availability
Joint
bottom joint: ST/NS14/20
joint: ST/NS top14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A891420
Related Categories
General description
With an inner drip joint at the bottom and an outer joint at the top.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)