Key Documents
SYNA251914
Synthware enlarging adapter
top joint: ST/NS 19/22, bottom joint: ST/NS 14/20
Synonym(s):
Synthware A251914, glass connecting adapter
Select a Size
1 EA
€27.50
€27.50
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€27.50
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
€27.50
Please contact Customer Service for Availability
Joint
bottom joint: ST/NS14/20
joint: ST/NS top19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A251914
Related Categories
General description
Synthware enlarging adapter is used for connecting dissimilar standard taper joint sizes. The top joint (19/22) is larger that the bottom joint (14/20).
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)