SLW3775/02M
Pyrex® Chromatography column, plain, length 355 mm, stem O.D. 8 mm
Synonym(s):
Pyrex, SciLabware 3775/02M
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
3775/02M
SciLabware
L
355 mm , excluding stem
body O.D.
14 mm
stem L
45 mm
stem O.D.
8 mm
pore size
Grade 1 porosity
General description
- Simple and economical column - ideal for educational use
- With chemically resistant glass sintered disc - porosity 1
- Manufactured from Pyrex borosilicate glass
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service