Key Documents
SLW1060/18D
Pyrex® Watch glasses
nominal diameter 125 mm
Synonym(s):
Pyrex, SciLabware 1060/18D
Select a Size
10 EA
€257.00
€257.00
Please contact Customer Service for Availability
Select a Size
Change View
10 EA
€257.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
€257.00
Please contact Customer Service for Availability
packaging
pkg of 10
manufacturer/tradename
1060/14D
SciLabware
nominal diameter
125 mm
General description
Watch glasses
  • For evaporation work and covering heated beakers
  • Manufactured from heat resisting Pyrex borosilicate glass
  • Fire-polished edges to reduce chipping and breakage
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)