Key Documents
SLW1060/04D
Pyrex® Watch glasses
nominal diameter 40 mm
Synonym(s):
Pyrex, SciLabware 1060/04D
Select a Size
10 EA
€176.00
€176.00
Please contact Customer Service for Availability
Select a Size
Change View
10 EA
€176.00
About This Item
UNSPSC Code:
41121801
NACRES:
NB.43
€176.00
Please contact Customer Service for Availability
packaging
pkg of 10
manufacturer/tradename
1060/04D
SciLabware
nominal diameter
40 mm
General description
Watch glasses
  • For evaporation work and covering heated beakers
  • Manufactured from heat resisting Pyrex borosilicate glass
  • Fire-polished edges to reduce chipping and breakage
Features and Benefits
  • Complies with DIN 12341
  • Manufactured from borosilicate glass 3.3
  • Heat resistant
  • Fire-polished edges to reduce chipping and breakage
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)