Documents
SLW1000/30D
Pyrex® Griffin beakers, low form, with printed trace code
3000 mL
Synonym(s):
Pyrex, SciLabware 1000/30D
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
material
borosilicate glass
white graduation
feature
low form
with printed trace code
manufacturer/tradename
1000/30D
SciLabware
H × diam.
210 mm × 150 mm
capacity
3000 mL
General description
  • Complies with ISO 3819 and DIN 12331
  • Manufactured from Pyrex borosilicate glass
  • Excellent thermal performance
  • High resistance to chemical attack
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • White graduations and marking spot
Features and Benefits
• Complies with ISO 3819 and DIN 12331• Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)• Excellent thermal performance• High resistance to chemical attack• Printed with Trace Code for downloadable batch certificate• White graduations and marking spot• Graduations are approximate• 1000/38D is supplementary to ISO range
Other Notes
Browse all glass beakers
Legal Information
PYREX is a registered trademark of Corning, Inc.