Skip to Content
Merck
All Photos(1)

Key Documents

SIK4116-20

Sigma-Aldrich

12,12,13,13,14,14,15,15,16,16,17,17,17-Tridecafluoroheptadecyltrimethoxysilane

95%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€158.00
1 G
€479.00

€158.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€158.00
1 G
€479.00

About This Item

Empirical Formula (Hill Notation):
C20H31F13O3Si
Molecular Weight:
594.52
UNSPSC Code:
12352103
NACRES:
NA.23

€158.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

95%

form

liquid

shelf life

3 years under the recommended conditions

Application

These products are used to form water repellent surface coating and/or lubricated surfaces on silicon, silicon oxide, etc.
The fluorinated carbon gives the omniphobic properties to the surface. The coated surface is more water repellent when a product having more fluorinated carbons is used.
The silane group provide a linker to covalently bond to silicon and silicon oxide surfaces.
The methoxy groups are more reactive than the ethoxy ones, but they release methanol instead of ethanol for ethoxy groups.

Legal Information

Product of SiKEMIA

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service