Skip to Content
Merck
All Photos(2)

Key Documents

S0764

Sigma-Aldrich

Sodium thiophosphate tribasic hydrate

≥90%

Synonym(s):

Sodium phosphorothioate hydrate, Trisodium thiophosphate hydrate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€160.00

€160.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€160.00

About This Item

Linear Formula:
Na3PO3S · xH2O
CAS Number:
Molecular Weight:
180.01 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

€160.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥90%

form

powder

storage temp.

2-8°C

SMILES string

O.[Na].OP(O)(S)=O

InChI

1S/3Na.H3O3PS.H2O/c;;;1-4(2,3)5;/h;;;(H3,1,2,3,5);1H2/q3*+1;;/p-3

InChI key

VYYYFZBRAKXIPU-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Chen et al.
Journal of cellular and molecular medicine, 16(7), 1447-1460 (2011-08-23)
Cyclic AMP (cAMP) is the archetypal smooth muscle relaxant, mediating the effects of many hormones and drugs. However, recently PGI(2) , acting via cAMP/PKA, was found to increase contraction-associated protein expression in myometrial cells and to promote oxytocin-driven myometrial contractility.
Alberto Balderas et al.
Neurochemistry international, 73, 139-145 (2013-11-12)
Glutamate, the major excitatory amino acid, activates a wide variety of signal transduction cascades. Ionotropic and metabotropic glutamate receptors are critically involved in long-term synaptic changes, although recent findings suggest that the electrogenic Na(+)-dependent glutamate transporters, responsible for its removal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service