RNI00087
Ritter Iminopyridine Ferrous Chloride Polymerization Catalyst
Synonym(s):
Iron, dichloro[5′-phenyl-N-[(2-pyridinyl-N)methylene][1,1′:3′,1′′-terphenyl]-2′-amine-N]-
About This Item
Recommended Products
form
solid
reaction suitability
core: iron
reagent type: catalyst
greener alternative product characteristics
Catalysis
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
greener alternative category
SMILES string
Cl[Fe]1(Cl)[N](C2=C(C3=CC=CC=C3)C=C(C4=CC=CC=C4)C=C2C5=CC=CC=C5)=CC6=CC=CC=[N]61
InChI
1S/C30H22N2.2ClH.Fe/c1-4-12-23(13-5-1)26-20-28(24-14-6-2-7-15-24)30(32-22-27-18-10-11-19-31-27)29(21-26)25-16-8-3-9-17-25;;;/h1-22H;2*1H;/q;;;+2/p-2/b32-22+;;;
InChI key
DYJUXFJEVQHNMN-KFIQQVOFSA-L
Related Categories
General description
Other Notes
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
The Ritter lab currently focuses on fluorination chemistry for late-stage functionalization of complex natural and unnatural products. PhenoFluor™ has been developed as a general reagent for the selective, predictable, direct deoxyfluorination of complex alcohols and phenols.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service