Skip to Content
Merck
All Photos(2)

Documents

R856

Sigma-Aldrich

Resorcinol monoacetate

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C6H4-3-(OH)
CAS Number:
Molecular Weight:
152.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

refractive index

n20/D 1.537 (lit.)

bp

283 °C (lit.)

density

1.223 g/mL at 25 °C (lit.)

SMILES string

CC(=O)Oc1cccc(O)c1

InChI

1S/C8H8O3/c1-6(9)11-8-4-2-3-7(10)5-8/h2-5,10H,1H3

InChI key

ZZPKZRHERLGEKA-UHFFFAOYSA-N

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federal register, 75(42), 9767-9777 (2010-04-14)
We, the Food and Drug Administration (FDA), are issuing this final rule to include benzoyl peroxide as a generally recognized as safe and effective (GRASE) active ingredient in over-the-counter (OTC) topical acne drug products. In addition, this final rule includes
S Gunasekaran et al.
Biomaterials, artificial cells, and artificial organs, 16(4), 771-783 (1988-01-01)
Collagen and gelatin containing biomaterials are relatively more susceptible to bacterial infection. Systemic administration or local delivery of antibiotics after implantation does not seem to solve the problem either effectively or easily. Antibiotics may be incorporated in the implant; but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service