Skip to Content
Merck
All Photos(1)

Key Documents

QBD11200

Sigma-Aldrich

Bromoacetamido-dPEG ®4-TFP ester

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

1000 MG
€357.00

€357.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1000 MG
€357.00

About This Item

Empirical Formula (Hill Notation):
C19H24BrF4NO7
CAS Number:
Molecular Weight:
534.29
UNSPSC Code:
12352101
NACRES:
NA.22

€357.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reactivity: thiol reactive
reagent type: cross-linking reagent
reactivity: amine reactive

functional group

bromo
ester

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

BrCC(NCCOCCOCCOCCOCCC(OC1=C(F)C(F)=CC(F)=C1F)=O)=O

InChI key

KMXNKPMXZFZZAS-UHFFFAOYSA-N

Features and Benefits

Bromoacetamido-dPEG®₄ -TFP ester can be used to conjugate two different proteins or an antibody with a payload by reaction of the bromoacetamido with a free thiol and reaction of the TFP ester with a primary amine.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service