Skip to Content
Merck
All Photos(1)

Key Documents

QBD11086

Sigma-Aldrich

m-dPEG ®13-NHS ester

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H59NO17
Molecular Weight:
729.81
MDL number:
UNSPSC Code:
51171641
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: chemical modification reagent
reagent type: cross-linking reagent
reactivity: amine reactive

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

N1(C(=O)CCC1=O)OC(=O)CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC

InChI

1S/C32H59NO17/c1-37-6-7-39-10-11-41-14-15-43-18-19-45-22-23-47-26-27-49-29-28-48-25-24-46-21-20-44-17-16-42-13-12-40-9-8-38-5-4-32(36)50-33-30(34)2-3-31(33)35/h2-29H2,1H3

InChI key

KLSDLLFNPXNSEL-UHFFFAOYSA-N

Features and Benefits

m-dPEG®₁₃-NHS ester is an amine reactive hydrophilic dPEG modifier.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service