Skip to Content
Merck
All Photos(1)

Key Documents

QBD10722

Sigma-Aldrich

Hydroxy-dPEG®24-t-butyl ester

>90% (HPLC)

Synonym(s):

HO-PEG24-CO-Otbu, HO-PEG24-TBE, Hydroxy-PEG-t-butyl ester, Hydroxy-PEG24-t-butyl-ester, PEG1000-t-butyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€387.00

€387.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€387.00

About This Item

Empirical Formula (Hill Notation):
C55H110O27
Molecular Weight:
1203.45
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.22

€387.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>90% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O(CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(=O)OC(C)(C)C)CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI key

YMILISGQCISYOH-UHFFFAOYSA-N

Features and Benefits

Hydroxy-dPEG24-t-butyl ester is a versatile building block based upon a monodispersed polyethylene glycol (PEG) chain, comparable to a polymeric PEG1000. The single molecular weight, discrete-length PEG (dPEG) chain is 86 atoms (88.5 Å) long. A primary alcohol group terminates one end of the molecule. The opposite end is a propionic acid group protected as the tert-butyl ester. The terminal hydroxy group can be functionalized with various reactive groups to permit direct modification of surfaces, small molecules, or biomolecules. The tert-butyl ester deprotects with trifluoroacetic acid (TFA), exposing the terminal propionic acid, which can then be activated as the NHS or TFP ester or directly coupled to an amine using a suitable carbodiimide.

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service