Skip to Content
Merck
All Photos(1)

Key Documents

QBD10523

Sigma-Aldrich

Azido-dPEG®7-amine

Synonym(s):

23-Azido-3,6,9,12,15,18,21-heptaoxatricosan-1-amine, Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€298.00
1000 MG
€692.00

€298.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€298.00
1000 MG
€692.00

About This Item

Empirical Formula (Hill Notation):
C16H34N4O7
CAS Number:
Molecular Weight:
394.46
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

€298.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reaction type: click chemistry
reagent type: cross-linking reagent
reaction type: click chemistry

functional group

amine
azide

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

NCCOCCOCCOCCOCCOCCOCCOCCN=[N+]=[N-]

InChI

1S/C16H34N4O7/c17-1-3-21-5-7-23-9-11-25-13-15-27-16-14-26-12-10-24-8-6-22-4-2-19-20-18/h1-17H2

InChI key

VZKXLNRXAFTBOW-UHFFFAOYSA-N

Features and Benefits

The azido-dPEG®7-amine contains an azide function on one end of a single molecular weight dPEG® spacer (28.8 Å) and a reactive group on the other end of the spacer. The dPEG® spacer is hydrophilic and non-immunogenic and improves the water solubility of the target molecule while reducing the immunogenicity and increasing the hydrodynamic volume of the target. The amine group reacts with acids and active esters. The azide group reacts with an alkyne in the well-known click chemistry reaction. The click chemistry reaction proceeds by copper(I) or ruthenium catalysis or in a strain-catalyzed reaction with certain types of alkyne partners.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service