Skip to Content
Merck
All Photos(1)

Key Documents

QBD10401

Sigma-Aldrich

NHS-dPEG®4-( m-dPEG®12)3-ester

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€302.00
1000 MG
€1,800.00

€302.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€302.00
1000 MG
€1,800.00

About This Item

Empirical Formula (Hill Notation):
C108H206N6O52
CAS Number:
Molecular Weight:
2420.80
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

€302.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: chemical modification reagent
reagent type: cross-linking reagent
reactivity: amine reactive

polymer architecture

shape: branched
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

N1(C(=O)CCC1=O)OC(=O)CCCC(=O)NCCOCCOCCOCCOCCC(=O)NC(COCCC(=O)NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)(COCCC(=O)NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)COCCC(=O)NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC

InChI

1S/C108H206N6O52/c1-123-25-28-131-41-44-139-55-58-145-67-70-151-79-82-157-91-94-160-88-85-154-76-73-148-64-61-142-52-49-136-38-33-128-22-14-110-101(116)9-18-163-97-108(113-104(119)12-17-126-31-36-134-47-48-135-37-32-127-21-13-109-100(115)5-4-6-107(122)166

InChI key

VOPFWBJWEJMWRQ-UHFFFAOYSA-N

Features and Benefits

NHS-dPEG®4-(m-dPEG12)3-ester is highly useful for protein and peptide modification to increase hydrodynamic volume and serum half-life and to greatly reduce the immunogenicity of the modified peptide or protein. These dPEG® products consist of a linear "tail" with a reactive group, a tris core to provide branching, and methoxy-terminated dPEG® branches. All of these dPEG® products are hydrophilic, non-immunogenic, single molecular weight compounds. N-hydroxysuccinimide (NHS) group reacts with free amines.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service