Key Documents
PL800SHELFI
Plas-Labs transport support cart
sliding shelf package - single station
Select a Size
1 EA
€2,220.00
€2,220.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€2,220.00
About This Item
UNSPSC Code:
41123300
NACRES:
NB.35
€2,220.00
Please contact Customer Service for Availability
H × W × D
18 in. × 18 in. × 10 in.
General description
Suspended sliding shelf for single operator units. Culture plate capacity is about 340. Fabricated of .375 in. thick clear acrylic, with shelves spaced at 7 in. apart. The entire assembly is suspended from the chamber top to keep the floor area uncluttered.
All carts have anti-scuff casters including brakes and locks. Carts also feature a cushioning system that minimizes vibration, shocks, and wear and tear on chambers.
Choose from one of the most recent versions:
Certificates of Analysis (COA)