Skip to Content
Merck
All Photos(1)

Key Documents

P53209

Sigma-Aldrich

Propylene sulfide

≥96%

Synonym(s):

Methylthiirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6S
CAS Number:
Molecular Weight:
74.14
Beilstein:
383559
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥96%

refractive index

n20/D 1.475 (lit.)

bp

72-75 °C (lit.)

density

0.946 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC1CS1

InChI

1S/C3H6S/c1-3-2-4-3/h3H,2H2,1H3

InChI key

MBNVSWHUJDDZRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masamichi Yamanaka et al.
Chemical communications (Cambridge, England), (15)(15), 1690-1691 (2004-07-28)
Three chiral guests in a cylindrical capsule led to six diastereomeric complexes.
Cong-Duan Vo et al.
Macromolecular rapid communications, 34(2), 156-162 (2013-01-16)
We report for the first time the combination of ATRP and ring-opening episulfide polymerization as a means to synthesize polysulfide-based low-dispersity amphiphilic block copolymers. The most significant finding is the possibility to perform ATRP under mild conditions using poly(propylene sulfide)
J P Bearinger et al.
Nature materials, 2(4), 259-264 (2003-04-12)
Alkanethiolates have been widely used as chemisorbates to modify gold surfaces, in spite of their relatively poor oxidative stability. We introduce gold-chemisorbing block copolymers bearing an anchoring block of poly(propylene sulphide) (PPS), selected in the expectation of greater stability. These
Biomaterials: Stable antifouling surfaces.
Ralph G Nuzzo
Nature materials, 2(4), 207-208 (2003-04-12)
T Matsuda et al.
Cancer letters, 86(2), 229-234 (1994-11-11)
Two organosulfur compounds, methyl propyl disulfide (MPD) and propylene sulfide (PS) from garlic and onions, were studied for their modifying effects on hepatocarcinogenesis in the F344 rats. Modifying potential was scored by comparing the number and area per cm2 of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service