Skip to Content
Merck
All Photos(1)

Key Documents

P50609

Sigma-Aldrich

1,3-Propanedithiol

99%

Synonym(s):

1,3-Dimercaptopropane, Trimethylene dimercaptan

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HS(CH2)3SH
CAS Number:
Molecular Weight:
108.23
Beilstein:
1071197
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

vapor pressure

5 mmHg ( 37.7 °C)

Assay

99%

form

liquid

refractive index

n20/D 1.539 (lit.)

bp

169 °C (lit.)

mp

−79 °C (lit.)

density

1.078 g/mL at 25 °C (lit.)

SMILES string

SCCCS

InChI

1S/C3H8S2/c4-2-1-3-5/h4-5H,1-3H2

InChI key

ZJLMKPKYJBQJNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuchen Wang et al.
Advanced materials (Deerfield Beach, Fla.), 32(46), e2004270-e2004270 (2020-10-13)
Liquid crystal elastomers (LCEs) are of interest for applications such as soft robotics and shape-morphing devices. Among the different actuation mechanisms, light offers advantages such as spatial and local control of actuation via the photothermal effect. However, the unwanted aggregation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service