Skip to Content
Merck
All Photos(1)

Key Documents

O8305

Sigma-Aldrich

L-Ornithine hydrochloride

99%

Synonym(s):

(S)-(+)-2,5-Diaminopentanoic acid hydrochloride, (S)-2,5-Diaminopentanoic acid monohydrochloride, L-Ornithine monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)3CH(CO2H)(NH2) · HCl
CAS Number:
Molecular Weight:
168.62
Beilstein:
3625847
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

99%

optical activity

[α]24/D +23.6°, c = 4 in 6 M HCl

mp

245 °C (dec.) (lit.)

SMILES string

Cl.NCCC[C@H](N)C(O)=O

InChI

1S/C5H12N2O2.ClH/c6-3-1-2-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H/t4-;/m0./s1

InChI key

GGTYBZJRPHEQDG-WCCKRBBISA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tom Rimaux et al.
Research in microbiology, 164(3), 216-225 (2012-11-28)
Arginine conversion through the arginine deiminase (ADI) pathway is a common metabolic trait of Lactobacillus sakei which is ascribed to an arc operon and which inquisitively involves citrulline excretion and re-uptake. The aim of this study was to verify whether
Sherry Dadsetan et al.
Biochemical pharmacology, 85(1), 115-123 (2012-10-30)
Combined administration of ornithine and phenylacetate (OP) is proposed as a novel treatment of hyperammonemia and hepatic encephalopathy. Ornithine is believed to increase ammonia fixation into glutamine in muscle tissue and glutamine is subsequently thought to react with phenylacetate forming
Charles D Mills
Critical reviews in immunology, 32(6), 463-488 (2013-02-23)
The purpose of immunology is simple. Cure or prevent disease. M1/M2 is useful because it is simple. M1/M2 describes the two major and opposing activities of macrophages. M1 activity inhibits cell proliferation and causes tissue damage while M2 activity promotes
Irene Tsiapa et al.
Nuclear medicine and biology, 40(2), 262-272 (2012-12-15)
Radiolabeled RGD peptides that specifically target integrin α(ν)β(3) have great potential in early tumor detection through noninvasive monitoring of tumor angiogenesis. Based on previous findings of our group on radiopeptides containing positively charged aminoacids, we developed a new cyclic cRGDfK
Nicole Berthold et al.
Antimicrobial agents and chemotherapy, 57(1), 402-409 (2012-11-02)
Proline-rich antimicrobial peptides (PrAMPs) from insects and mammals have recently been evaluated for their pharmaceutical potential in treating systemic bacterial infections. Besides the native peptides, several shortened, modified, or even artificial sequences were highly effective in different murine infection models.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service