Skip to Content
Merck
All Photos(1)

Key Documents

O5209

Sigma-Aldrich

2-Octenoic acid

technical grade, 85%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€47.90

€47.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€47.90

About This Item

Linear Formula:
CH3(CH2)4CH=CHCO2H
CAS Number:
Molecular Weight:
142.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€47.90


Please contact Customer Service for Availability

Request a Bulk Order

grade

technical grade

description

predominantly trans

Assay

85%

refractive index

n20/D 1.4588 (lit.)

bp

154 °C/22 mmHg (lit.)

mp

5-6 °C (lit.)

density

0.944 g/mL at 25 °C (lit.)

SMILES string

CCCCC\C=C\C(O)=O

InChI

1S/C8H14O2/c1-2-3-4-5-6-7-8(9)10/h6-7H,2-5H2,1H3,(H,9,10)/b7-6+

InChI key

CWMPPVPFLSZGCY-VOTSOKGWSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service