Skip to Content
Merck
All Photos(1)

Key Documents

O4806

Sigma-Aldrich

1-Octene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH=CH2
CAS Number:
Molecular Weight:
112.21
Beilstein:
1734497
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.9 (vs air)

vapor pressure

36 mmHg ( 38 °C)

Assay

98%

form

liquid

autoignition temp.

446 °F

expl. lim.

3.9 %

refractive index

n20/D 1.408 (lit.)
n20/D 1.409 (lit.)

bp

122-123 °C (lit.)

mp

−101 °C (lit.)

density

0.715 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC=C

InChI

1S/C8H16/c1-3-5-7-8-6-4-2/h3H,1,4-8H2,2H3

InChI key

KWKAKUADMBZCLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Octene is a linear α-olefin mainly used as a comonomer in the synthesis of linear low-density polyethylene (LLDPE). Extensive work has been reported on the hydroformylation of 1-octene using various catalysts. It is also an important source to synthesize important petrochemical building blocks via epoxidation reaction.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immobilization of a rhodium carbene complex to an amphiphilic block copolymer for hydroformylation of 1-octene under aqueous two-phase conditions
Zarka, M Tobias and Bortenschlager, Martin and Wurst, Klaus and Nuyken, Oskar and Weberskirch, Ralf
Organometallics, 23(21), 4817-4820 (2004)
Gas-liquid-liquid reaction engineering: hydroformylation of 1-octene using a water soluble rhodium complex catalyst
Purwanto, P and Delmas, H
Catalysis Today, 24(1-2), 135-140 (1995)
Hydroperoxide oxidations catalyzed by metals. IV. Molybdenum hexacarbonyl catalyzed epoxidation of 1-octene
Baker III, TN and Mains, GJ and Sheng, MN and Zajacek, JG
The Journal of Organic Chemistry, 38(6), 1145-1148 (1973)
Catalytic epoxidation of 1-octene with diluted hydrogen peroxide. On the basic role of hydroxo complexes of platinum (II) and related species
Strukul, Giorgio and Michelin, Rino A
Journal of the American Chemical Society, 107(25), 7563-7569 (1985)
Ethylene tetramerization: a new route to produce 1-octene in exceptionally high selectivities
Bollmann A, et al.
Journal of the American Chemical Society, 126(45), 14712-14713 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service