Skip to Content
Merck
All Photos(1)

Key Documents

MT51343184

Mettler-Toledo electrolyte for reference electrodes

3 mol/L KCl Sat AgCl, volume 25 mL

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
€12.00

€12.00


Please contact Customer Service for Availability


Select a Size

Change View
25 ML
€12.00

About This Item

UNSPSC Code:
51191600
NACRES:
NB.79

€12.00


Please contact Customer Service for Availability

form

liquid

manufacturer/tradename

Mettler-Toledo 51343184

volume

25 mL

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service