Skip to Content
Merck
All Photos(1)

Key Documents

MT51343165

Mettler-Toledo InLab® Semi-Micro pH electrode

for measuring in small or narrow neck vessels

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115604
NACRES:
NB.79
Pricing and availability is not currently available.

material

glass membrane (A41)
glass shaft

description

Reference electrolyte XEROLYT EXTRA Polymer
Reference system ARGENTHAL
S7 connector
Without cable

manufacturer/tradename

Mettler-Toledo 51343165

temperature measuring range

0-100 °C

L × diam.

100 mm × 6 mm

pH range

0-12

General description

The InLab® Semi-Micro is a glass-body, semi-micro combination pH electrode with S7 screw head.
Designed for measuring in small or narrow neck vessels.

Designed with a shaft diameter of 6 mm the InLab® Semi-Micro, is able to measure pH in small or narrow neck vessels. Combination design avoids use of separate reference and indicating electrodes.

The InLab® Semi-Micro contains the latest in polymer electrolytes: XEROLYT EXTRA, which is characterized by high signal stability, a very short response time and high chemical resistance. Service and operation could not be simpler thanks to the polymer electrolyte and the open reference connection. Different cables can be connected to the S7 screw head allowing use with a wide range of meters.
  • 6 mm diameter shaft
  • Combination design
  • Detachable cable

Legal Information

ARGENTHAL is a trademark of Mettler-Toledo, Inc.
InLab is a registered trademark of Mettler-Toledo, Inc.
XEROLYT is a trademark of Mettler-Toledo, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service