Skip to Content
Merck
All Photos(1)

Key Documents

MT51302602

Mettler-Toledo SevenGo Duo pH/conductivity

SG23 field kit, InLab® Expert Pro-ISM and InLab® 738-ISM, cable L 2 m

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603
NACRES:
NB.61

description

Calibration Max. 3 points, 4 predefined buffer groups
Conductivity range 0.10 μS/cm ... 500 mS/cm (+/-0.5%)
Resistivity range 0.00 ... 100.00 MOhm*cm
Temperature range °C pH/mV: -5.0 ... 105/LF: -5.0 ... 105.0 (+/- 0.2)

manufacturer/tradename

Mettler-Toledo 51302602

TDS measuring range

0.1 mg/L (min)
300 g/L (max)

mV measuring range

-1999-1999, accuracy: ±1

cable L

2 m

battery power

4 AA (included in standard delivery)

pH range

0-14

Looking for similar products? Visit Product Comparison Guide

General description

Includes meter, InLab® Expert Pro-ISM and InLab® 738-ISM (both IP67, 2 m cable), rubber holster, calibration sachets, sample bottles, 2-electrode clip and Field Compact Case
SevenGo Duo combines user-friendliness, quality and reliable measurements. The meter is designed so that a single keystroke triggers a calibration or measurement or allows accessing stored results or system settings. The kit versions are also well suited for outdoor use or use in the production area because both the meter and the sensors are watertight (IP67).
The revolutionary intelligent sensor management system (ISM®) offers the user an even greater level of security. The calibration data is stored on the chip in the sensor and is used automatically when the sensor is connected to the meter. This ensures that the meter always uses the latest calibration data.

Features and Benefits
Routine pH/Conductivity Meter (IP67)
Portable dual-channel meter for the determination of pH, mV, conductivity, TDS (total dissolved solids), salinity and specific resistance

3-point pH calibration with selection from 4 predefined and one user-defined buffer groups

Data memory for 99 GLP data records

Intelligent sensor management (ISM)

Outstanding operation due to high-contrast display with large figures

Reproducible results due to automatic end point detection, automatic buffer detection and temperature compensation

Rugged watertight housing (IP67)

Legal Information

ISM is a registered trademark of Mettler-Toledo, Inc.
InLab is a registered trademark of Mettler-Toledo, Inc.
SevenGo is a trademark of Mettler-Toledo, Inc.
SevenGo Duo is a trademark of Mettler-Toledo, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service