MT30207972
Mettler Toledo Seven2Go™ Pro DO meter S9
Field Kit, optical
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
connector mini-LTW (IP67)
interface: micro-USB
pH/ion/ORP meters
feature
Field Kit, optical
manufacturer/tradename
Mettler-Toledo 30207972
dissolved oxygen measuring range
0.00-50.0 mg/L (ppm)
pressure measuring range
500-1100 (mbar)
saturation measuring range
0.0-500% (DO)
temperature measuring range
0-50 °C
battery power
4 x 1.5 V AA (or 1.3 V NiMH accumulators)
Looking for similar products? Visit Product Comparison Guide
General description
Simple and reliable, attractively designed and robust. The Seven2Go portables are engineered to provide users with fast quality data, one-handed operation, and durability, making them ideally suited for mobile applications in the laboratory, at-line, or outdoors.
Simple Menu – Intuitive Operation:
T-Pad – Comfortable, Fast Navigation:
ISM® Ready – Reduced Chances for Error:
IP67 – Weatherproof and Robust Design:
12 Languages – No more Confusion:
Sunlight-proof – Always Brilliant Readability:
StatusLight – Easy Readiness Check:
Simple Menu – Intuitive Operation:
- Seven2Go′s new intuitive menu guarantees out-of-the-box operation for anyone. Users no longer need to study lengthy operating instructions
T-Pad – Comfortable, Fast Navigation:
- Seven2Go′s new T-Pad improves the speed with which you move through menus. The smart button arrangement allows for faster navigation that provide you shorter time to actual measurement
ISM® Ready – Reduced Chances for Error:
- Upon connection of any InLab ISM (Intelligent Sensor Management) sensor, the instrument automatically recognizes the sensor and stores sensor ID, serial number, and last calibration data
IP67 – Weatherproof and Robust Design:
- Thanks to its IP67 protection and drop test resistance, the new Seven2Go portables are able to withstand harsh and demanding environments
12 Languages – No more Confusion:
- The Seven2Go supports 12 languages and offers a user interface that uses complete words and sentences. The lack of cryptic expressions simplify operation and reduces strain on the user
Sunlight-proof – Always Brilliant Readability:
- The screen′s contrast is enhanced by direct sunlight. This guarantees an excellent readability even under bright and sunny conditions
StatusLight – Easy Readiness Check:
- Thanks to the new StatusLight you are instantly aware of how the meter is doing. An easy to understand traffic light concept shows if your meter is ready or not
Application
Fast results in aqueous samples on the go
Features and Benefits
Includes: Meter, LabX direct pH, InLab OptiOx with protective guard, USB cable, electrode clip, base stand, instructions, quick guide, test certificate, declaration of conformity, batteries, bottles for calibration, zero oxygen tablets and uGO carrying case
Legal Information
ISM is a registered trademark of Mettler-Toledo, Inc.
Seven2Go is a trademark of Mettler-Toledo, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service