Skip to Content
Merck
All Photos(1)

Key Documents

M86006

Sigma-Aldrich

Methyl 3,4,5-trimethoxybenzoate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€31.40

€31.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€31.40

About This Item

Linear Formula:
(CH3O)3C6H2CO2CH3
CAS Number:
Molecular Weight:
226.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€31.40


Please contact Customer Service for Availability

Request a Bulk Order

Assay

98%

form

solid

bp

274-275 °C (lit.)

mp

82-84 °C (lit.)

SMILES string

COC(=O)c1cc(OC)c(OC)c(OC)c1

InChI

1S/C11H14O5/c1-13-8-5-7(11(12)16-4)6-9(14-2)10(8)15-3/h5-6H,1-4H3

InChI key

KACHFMOHOPLTNX-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service