Skip to Content
Merck
All Photos(1)

Key Documents

M57405

Sigma-Aldrich

Menadione

98%

Synonym(s):

2-Methyl-1,4-naphthoquinone, Vitamin K3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8O2
CAS Number:
Molecular Weight:
172.18
Beilstein:
1908453
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

synthetic

Assay

98%

form

powder

technique(s)

HPLC: suitable

color

yellow to yellow-green

mp

105-107 °C (lit.)

SMILES string

CC1=CC(=O)c2ccccc2C1=O

InChI

1S/C11H8O2/c1-7-6-10(12)8-4-2-3-5-9(8)11(7)13/h2-6H,1H3

InChI key

MJVAVZPDRWSRRC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dipankar Koley et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(29), 11522-11527 (2012-06-09)
Oxidative stress induced in live HeLa cells by menadione (2-methyl-1,4-napthaquinone) was studied in real time by scanning electrochemical microscopy (SECM). The hydrophobic molecule menadione diffuses through a living cell membrane where it is toxic to the cell. However, in the
Juan Lucas Argueso et al.
Genome research, 19(12), 2258-2270 (2009-10-09)
Bioethanol is a biofuel produced mainly from the fermentation of carbohydrates derived from agricultural feedstocks by the yeast Saccharomyces cerevisiae. One of the most widely adopted strains is PE-2, a heterothallic diploid naturally adapted to the sugar cane fermentation process
H H Thijssen et al.
The British journal of nutrition, 75(1), 121-127 (1996-01-01)
We measured the vitamin K status in postmortem human tissues (brain, heart, kidney, liver, lung, pancreas) to see if there is a tissue-specific distribution pattern. Phylloquinone (K1) was recovered in all tissues with relatively high levels in liver, heart and
Y Usui et al.
The American journal of clinical nutrition, 51(5), 846-852 (1990-05-01)
Vitamin K deficiency has been reported in patients who were treated with antibiotics and placed on poor diets after surgery. High-performance liquid chromatography (HPLC) was used to study the influence of dietary intake on vitamin K concentrations in surgical patients
B Wermuth et al.
Biochemical pharmacology, 35(8), 1277-1282 (1986-04-15)
Enzymes catalyzing the two-electron reduction of quinones to hydroquinones are thought to protect the cell against quinone-induced oxidative stress. Using menadione as a substrate, carbonyl reductase, a cytosolic, monomeric oxidoreductase of broad specificity for carbonyl compounds, was found to be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service