Skip to Content
Merck
All Photos(1)

Documents

M26593

Sigma-Aldrich

2′-Methylacetophenone

98%

Synonym(s):

1-(2-Methylphenyl)ethanone, 1-(2-Tolyl)ethanone, 2-Acetyltoluene, o-Methylacetophenone, Methyl o-tolyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4COCH3
CAS Number:
Molecular Weight:
134.18
Beilstein:
907005
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.5318 (lit.)

bp

214 °C (lit.)

density

1.026 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1ccccc1C

InChI

1S/C9H10O/c1-7-5-3-4-6-9(7)8(2)10/h3-6H,1-2H3

InChI key

YXWWHNCQZBVZPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service