M2004
(S)-(+)-Mandelic acid
ReagentPlus®, ≥99%
Synonym(s):
(S)-α-Hydroxyphenylacetic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5CH(OH)CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein:
2208678
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
product line
ReagentPlus®
Assay
≥99%
optical activity
[α]20/D +154°, c = 2.8 in H2O
optical purity
ee: 99% (GLC)
mp
131-134 °C (lit.)
SMILES string
O[C@H](C(O)=O)c1ccccc1
InChI
1S/C8H8O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7,9H,(H,10,11)/t7-/m0/s1
InChI key
IWYDHOAUDWTVEP-ZETCQYMHSA-N
Looking for similar products? Visit Product Comparison Guide
General description
N-Methyl-D-glucamine (Meglumine) is a sorbitol derivative in which one of the hydroxyl groups has been replaced with a methylamino group. Meglumine is widely used with iodinated organic compounds as a contrast media.
Application
N-Methyl-D-glucamine can be used as a co-crystal conformer as well as a salt-forming agent due to its hydrophilic nature and ability to modify the crystalline structure of the drug molecule to improve solubility.
It is also used to functionalize polymers and resins to develop chelating agents for the removal of boron and arsenic from wastewater.
It is also used to functionalize polymers and resins to develop chelating agents for the removal of boron and arsenic from wastewater.
A versatile reagent used in the resolution of racemates and the preparation of amides.
Legal Information
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
>374.0 °F
Flash Point(C)
> 190 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The Journal of Organic Chemistry, 58, 2313-2313 (1993)
Tetrahedron, 50, 5049-5049 (1994)
Melanie Balbach et al.
Frontiers in cell and developmental biology, 8, 572735-572735 (2020-09-29)
Mammalian oocytes are enveloped by the zona pellucida (ZP), an extracellular matrix of glycoproteins. In sperm, stimulation with ZP proteins evokes a rapid Ca2+ influx via the sperm-specific, pH-sensitive Ca2+ channel CatSper. However, the physiological role and molecular mechanisms underlying
Synthetic Communications, 23, 2761-2761 (1993)
Janine Obendorf et al.
Stem cell research & therapy, 11(1), 525-525 (2020-12-11)
Mesenchymal stem cells (MSCs) were shown to harbor therapeutic potential in models of respiratory diseases, such as bronchopulmonary dysplasia (BPD), the most common sequel of preterm birth. In these studies, cells or animals were challenged with hyperoxia or other injury-inducing
Chromatograms
suitable for GCsuitable for GCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service