Skip to Content
Merck
All Photos(1)

Key Documents

L080000-HH

Sigma-Aldrich

DCI Activator

0.25M

Synonym(s):

DCI activator solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

type

for DNA synthesis

product line

Proligo Reagents

packaging

pkg of 4 × 2.5 L (L080250-04)
pkg of 1 × 25 L (L082500-01)
pkg of 4 × 4 L (L080400-04)

concentration

29-32 g/L (Dicyanoimidazole, titration)

impurities

≤30 ppm water content (Karl Fischer)

color

clear

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Dicyanoimidazole / Acetonitrile 3/100 (w/v)

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

41.0 °F

Flash Point(C)

5 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service