Skip to Content
Merck
All Photos(1)

Key Documents

L021000

Sigma-Aldrich

DCA Deblock

with Dichloromethane

Sign Into View Organizational & Contract Pricing

Select a Size

4 X 2.5 L
€630.00

€630.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
4 X 2.5 L
€630.00

About This Item

UNSPSC Code:
41116004
NACRES:
NA.21

€630.00


Please contact Customer Service for Availability

Request a Bulk Order

type

for DNA synthesis
for DNA synthesis

product line

Proligo Reagents

form

liquid

storage condition

dry at room temperature

concentration

42.2-51.6 g/L (DCA content, titration)

impurities

≤200 ppm water content (Karl Fischer)

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

Deblock solution contains dichloroacetic acid (DCA) in dichloromethane that assists in the removal of the dimethoxytrityl (DMT) protecting group from the 5′hydroxyl moiety of nucleotides already incorporated into the growing nucleic acid, before the addition of the next phosphoramidite. Removal of the DMT allows the unprotected 5′hydroxyl moiety to react with a new phosphoramidite in a subsequent extension reaction.

Preparation Note

Dicloromethane / Dichloroacetic acid 97/3 (v/v)

Other Notes

Deblock solution containing dichloroacetic acid (DCA) in dichloromethane removes the dimethoxytrityl (DMT) protecting group from the 5′hydroxyl moiety of nucleotides already incorporated into the growing nucleic acid, prior to the addition of the next phosphoramidite. Removal of the DMT allows the unprotected 5′hydroxyl moiety to react with a new phosphoramidite in a subsequent extension reaction.

Signal Word

Danger

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service